missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Glutaryl Chloride 98.0+%, TCI America™
Supplier: TCI America E045125G
Specifications
Ethyl Glutaryl Chloride | |
Yellow | |
C7H11ClO3 | |
25 g | |
ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester | |
CCOC(=O)CCCC(=O)Cl | |
178.612 | |
178.61 | |
Liquid |
5205-39-0 | |
108°C | |
MFCD00013659 | |
3265 | |
KKJAQUGGQMCNJY-UHFFFAOYSA-N | |
ethyl 5-chloro-5-oxopentanoate | |
78881 | |
≥98.0% (T) |
Chemical Identifiers
5205-39-0 | |
178.612 | |
KKJAQUGGQMCNJY-UHFFFAOYSA-N | |
78881 | |
CCOC(=O)CCCC(=O)Cl |
C7H11ClO3 | |
MFCD00013659 | |
ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester | |
ethyl 5-chloro-5-oxopentanoate |
Safety and Handling
TSCA : No