Learn More
Ethyl formate, 98+%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150675000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl formate | |
-80.0°C | |
54.0°C | |
Authentic | |
Glass bottle | |
1.3580 to 1.3610 | |
MFCD00003294 | |
02, 19 | |
0.917 | |
areginal, formic acid, ethyl ester, ethyl methanoate, formic acid ethyl ester, aethylformiat, ethyl formic ester, ethylformate, ethylformiaat, formic ether, ethylformiat | |
WBJINCZRORDGAQ-UHFFFAOYSA-N | |
ethyl formate | |
8025 | |
CHEBI:52342 | |
98+% | |
Liquid |
109-94-4 | |
0.9170g/mL | |
−20°C | |
98% min. (GC) | |
C3H6O2 | |
HCO2C2H5 | |
500 mL | |
01,380; 02,197; 04,233; 12,226 | |
15, 3862 | |
Solubility in water: 110g/L (18°C). Other solubilities: freely soluble in ethylacetaat, isobutylacetaat,, butylacetaat, aceton, methylisobutylketon and, methylethylketon, miscible with benzene, ether and alcohol | |
CCOC=O | |
74.08 | |
0.4 mPa.s (20°C) | |
74.08 | |
Pure |
Chemical Identifiers
109-94-4 | |
74.08 | |
WBJINCZRORDGAQ-UHFFFAOYSA-N | |
8025 | |
ethyl formate |
C3H6O2 | |
MFCD00003294 | |
areginal, formic acid, ethyl ester, ethyl methanoate, formic acid ethyl ester, aethylformiat, ethyl formic ester, ethylformate, ethylformiaat, formic ether, ethylformiat | |
CHEBI:52342 | |
CCOC=O |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a POISON CENTER or doctor/physician if you
GHS Signal Word: Danger
EINECSNumber : 203-721-
RUO – Research Use Only