missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl formate, 97%

$35.55 - $35.55

Chemical Identifiers

CAS 109-94-4
Molecular Formula C3H6O2
Linear Formula HCO2CH2CH3
Formula Weight 74.08
MDL Number MFCD00003294
UN Number UN1190
Beilstein 906769
Merck Index 14,3807
Synonym Formic acid ethyl ester
Chemical Name or Material Ethyl formate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA111130F SDS Alfa Aesar™
A111130F
2500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA11113AE SDS Alfa Aesar™
A11113AE
100mL Each for $35.55
AAA11113AP SDS Alfa Aesar
A11113AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Ethyl formate is used as a solvent for cellulose nitrate, cellulose acetate, greases and oils. It is also used as a flavor for lemonade and essences. It is involved in the fumigation of dried fruits, as an alternative to methyl bromide.

Solubility
Miscible with water, benzene, alcohol, ether, acetone and most organic solvents.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing agents.
Description & Specifications

Chemical Identifiers

109-94-4
HCO2CH2CH3
MFCD00003294
906769
Formic acid ethyl ester
C3H6O2
74.08
UN1190
14,3807
Ethyl formate

Specifications

109-94-4
Liquid
C3H6O2
74.08
UN1190
14,3807
52° to 54°C
4.1
−19°C (−2°F)
97%
Fruit-like
97%
HCO2CH2CH3
MFCD00003294
906769
Formic acid ethyl ester
−81°C
2500mL
Ethyl formate

Safety and Handling

GHS H Statement
H225-H302-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P262-P243-P305+P351+P338-P403
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not get in eyes, on skin, or on clothing.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL FORMATE

RTECSNumber : LQ8400000

TSCA : TSCA

Recommended Storage : 2°C to 8°C