missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Fluoroacetate 98.0+%, TCI America™
Supplier: TCI America F003025ML
Specifications
Ethyl Fluoroacetate | |
Yellow | |
C4H7FO2 | |
25 mL | |
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester | |
CCOC(=O)CF | |
106.096 | |
106.10 | |
Liquid |
459-72-3 | |
117°C | |
MFCD00000450 | |
2929 | |
VCYZVXRKYPKDQB-UHFFFAOYSA-N | |
ethyl 2-fluoroacetate | |
9988 | |
≥98.0% (GC) |
Chemical Identifiers
459-72-3 | |
106.096 | |
VCYZVXRKYPKDQB-UHFFFAOYSA-N | |
9988 | |
CCOC(=O)CF |
C4H7FO2 | |
MFCD00000450 | |
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester | |
ethyl 2-fluoroacetate |
Safety and Handling
EINECSNumber : (2)-3837
RTECSNumber : AH7175000
TSCA : Yes