Learn More
Ethyl (ethoxymethylene)cyanoacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1236630
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl (ethoxymethylene)cyanoacetate | |
47°C to 52°C | |
130°C (266°F) | |
MFCD00009136 | |
UN3439 | |
ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate | |
CCO\C=C(\C#N)C(=O)OCC | |
169.18 | |
169.18 |
94-05-3 | |
190°C to 191°C (30 mmHg) | |
C8H11NO3 | |
250 g | |
878814 | |
KTMGNAIGXYODKQ-SREVYHEPSA-N | |
ethyl (E)-2-cyano-3-ethoxyprop-2-enoate | |
1715183 | |
98% |
Chemical Identifiers
94-05-3 | |
169.18 | |
KTMGNAIGXYODKQ-SREVYHEPSA-N | |
1715183 | |
CCO\C=C(\C#N)C(=O)OCC |
C8H11NO3 | |
MFCD00009136 | |
ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate | |
ethyl (E)-2-cyano-3-ethoxyprop-2-enoate |
Safety and Handling
GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H318-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 202-299-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only