missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Ethoxyacetate 95.0+%, TCI America™
$84.60 - $252.17
Chemical Identifiers
CAS | 817-95-8 |
---|---|
Molecular Formula | C6H12O3 |
Molecular Weight (g/mol) | 132.159 |
MDL Number | MFCD00040492 |
InChI Key | CKSRFHWWBKRUKA-UHFFFAOYSA-N |
Synonym | ethyl ethoxyacetate, ethoxyacetic acid ethyl ester, acetic acid, ethoxy-, ethyl ester, ethoxy-acetic acid ethyl ester, ethoxyacetic acid, ethyl ester, acetic acid, 2-ethoxy-, ethyl ester, diethyl glycolate, ethyl ethoxylacetate, acmc-20akxi |
PubChem CID | 69955 |
IUPAC Name | ethyl 2-ethoxyacetate |
SMILES | CCOCC(=O)OCC |
Chemical Identifiers
817-95-8 | |
132.159 | |
CKSRFHWWBKRUKA-UHFFFAOYSA-N | |
69955 | |
CCOCC(=O)OCC |
C6H12O3 | |
MFCD00040492 | |
ethyl ethoxyacetate, ethoxyacetic acid ethyl ester, acetic acid, ethoxy-, ethyl ester, ethoxy-acetic acid ethyl ester, ethoxyacetic acid, ethyl ester, acetic acid, 2-ethoxy-, ethyl ester, diethyl glycolate, ethyl ethoxylacetate, acmc-20akxi | |
ethyl 2-ethoxyacetate |
Specifications
817-95-8 | |
158°C | |
MFCD00040492 | |
3272 | |
CKSRFHWWBKRUKA-UHFFFAOYSA-N | |
ethyl 2-ethoxyacetate | |
69955 | |
≥95.0% (GC) | |
Ethyl Ethoxyacetate |
Yellow | |
C6H12O3 | |
5 g | |
ethyl ethoxyacetate, ethoxyacetic acid ethyl ester, acetic acid, ethoxy-, ethyl ester, ethoxy-acetic acid ethyl ester, ethoxyacetic acid, ethyl ester, acetic acid, 2-ethoxy-, ethyl ester, diethyl glycolate, ethyl ethoxylacetate, acmc-20akxi | |
CCOCC(=O)OCC | |
132.159 | |
132.16 | |
Liquid |
Safety and Handling
TSCA : Yes