missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Ether, Anhydrous, 99.0% min., ACS, EMD Millipore

Manufacturer:  Millipore Sigma EX01909

 View more versions of this product

Catalog No. MEX01909



Specifications

Specifications

Ethyl Ether, Anhydrous
60-29-7
C4H10O
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
Glass Bottle
1L
For ACS analysis
99.0% min.
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.123
CHEBI:35702
Liquid
Colorless

Chemical Identifiers

60-29-7
74.123
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC
C4H10O
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
Documents