missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Ether ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 60-29-7
Molecular Formula (C2H5)2O
Formula Weight 74.12
Chemical Name or Material Ethyl Ether
Grade ACS Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK0848212 Avantor Performance
084812
Al Bottle 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK0850212 Avantor Performance
085012
Al Bottle 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK0850500 Avantor Performance
085010
Al Bottle 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

60-29-7
74.12
ACS Reagent Grade
(C2H5)2O
Ethyl Ether

Specifications

74.12
60-29-7
Al Bottle
ACS Reagent Grade
2.5kg
(C2H5)2O
Ethyl Ether