Please login to your online account to display your discounted pricing

Ethyl Ether ACS AR, Macron Fine Chemicals™


macron

Quantity 500g
Packaging Al Bottle

Manufacturer: avantor performance  085010

 View more versions of this product

Catalog No. MK0850500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 74.12
Quantity 500g
Molecular Formula (C2H5)2O
CAS 60-29-7
Chemical Name or Material Ethyl Ether
Packaging Al Bottle

Specifications

Chemical Name or Material Ethyl Ether
CAS 60-29-7
Purity Grade ACS Reagent Grade
Molecular Formula (C2H5)2O
Formula Weight 74.12