missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl (E)-3-(1-pyrrolidinyl)crotonate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0046122
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl (E)-3-(1-pyrrolidinyl)crotonate | |
125°C to 126°C (0.8 mmHg) | |
1.541 | |
100 g | |
e-ethyl 3-pyrrolidin-1-yl but-2-enoate, ethyl e-3-1-pyrrolidinyl crotonate, 2e-3-1-pyrrolidinyl-2-butenoic acid ethyl ester, ethyl e-3-1-pyrrolidino crotonate, ethyl e-3-pyrrolidin-1-ylbut-2-enoate, e-3-pyrrolidin-1-yl-but-2-enoic acid ethyl ester, 2-butenoic acid, 3-1-pyrrolidinyl-, ethyl ester, e, ethyl 2e-3-pyrrolidin-1-yl but-2-enoate, ethyl 3-1-pyrrolidinyl crotonate | |
CCOC(=O)C=C(C)N1CCCC1 | |
183.251 | |
183.25 |
54716-02-8 | |
C10H17NO2 | |
MFCD00014097 | |
1310113 | |
MSOQKPXSIHLODG-CMDGGOBGSA-N | |
ethyl (E)-3-pyrrolidin-1-ylbut-2-enoate | |
736203 | |
97% |
Chemical Identifiers
54716-02-8 | |
183.251 | |
MSOQKPXSIHLODG-CMDGGOBGSA-N | |
736203 | |
CCOC(=O)C=C(C)N1CCCC1 |
C10H17NO2 | |
MFCD00014097 | |
e-ethyl 3-pyrrolidin-1-yl but-2-enoate, ethyl e-3-1-pyrrolidinyl crotonate, 2e-3-1-pyrrolidinyl-2-butenoic acid ethyl ester, ethyl e-3-1-pyrrolidino crotonate, ethyl e-3-pyrrolidin-1-ylbut-2-enoate, e-3-pyrrolidin-1-yl-but-2-enoic acid ethyl ester, 2-butenoic acid, 3-1-pyrrolidinyl-, ethyl ester, e, ethyl 2e-3-pyrrolidin-1-yl but-2-enoate, ethyl 3-1-pyrrolidinyl crotonate | |
ethyl (E)-3-pyrrolidin-1-ylbut-2-enoate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 259-303-3
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only