missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (E)-3-(1-pyrrolidinyl)crotonate, 97%

Manufacturer:  Alfa Aesar™ L0046114

 View more versions of this product

Catalog No. AAL0046114



Specifications

Specifications

Ethyl (E)-3-(1-pyrrolidinyl)crotonate
25g
C10H17NO2
1310113
MSOQKPXSIHLODG-CMDGGOBGSA-N
ethyl (E)-3-pyrrolidin-1-ylbut-2-enoate
736203
97%
125°C to 126°C (0.8mmHg)
54716-02-8
MFCD00014097
e-ethyl 3-pyrrolidin-1-yl but-2-enoate, ethyl e-3-1-pyrrolidinyl crotonate, 2e-3-1-pyrrolidinyl-2-butenoic acid ethyl ester, ethyl e-3-1-pyrrolidino crotonate, ethyl e-3-pyrrolidin-1-ylbut-2-enoate, e-3-pyrrolidin-1-yl-but-2-enoic acid ethyl ester, 2-butenoic acid, 3-1-pyrrolidinyl-, ethyl ester, e, ethyl 2e-3-pyrrolidin-1-yl but-2-enoate, ethyl e-3-1-pyrrolidino crotonate, ethyl 3-1-pyrrolidinyl crotonate
CCOC(=O)C=C(C)N1CCCC1
183.251
183.25

Chemical Identifiers

54716-02-8
183.251
MSOQKPXSIHLODG-CMDGGOBGSA-N
736203
CCOC(=O)C=C(C)N1CCCC1
C10H17NO2
MFCD00014097
e-ethyl 3-pyrrolidin-1-yl but-2-enoate, ethyl e-3-1-pyrrolidinyl crotonate, 2e-3-1-pyrrolidinyl-2-butenoic acid ethyl ester, ethyl e-3-1-pyrrolidino crotonate, ethyl e-3-pyrrolidin-1-ylbut-2-enoate, e-3-pyrrolidin-1-yl-but-2-enoic acid ethyl ester, 2-butenoic acid, 3-1-pyrrolidinyl-, ethyl ester, e, ethyl 2e-3-pyrrolidin-1-yl but-2-enoate, ethyl e-3-1-pyrrolidino crotonate, ethyl 3-1-pyrrolidinyl crotonate
ethyl (E)-3-pyrrolidin-1-ylbut-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : 2° to 8°C

Documents