missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl dodecanoate, 98%

Manufacturer:  Alfa Aesar™ A11188AP

 View more versions of this product

Catalog No. AAA11188AP


 

Specifications

Specifications

Ethyl dodecanoate
106-33-2
268°C to 270°C
98%
C14H28O2
1769671
Dodecanoic acid ethyl ester; Ethyl laurate
CCCCCCCCCCCC(=O)OCC
228.376
CHEBI:87427
500mL
-2°C
138°C (280°F)
Fruit-like
MFCD00015065
14,3818
MMXKVMNBHPAILY-UHFFFAOYSA-N
ethyl dodecanoate
7800
228.38

Chemical Identifiers

106-33-2
228.376
MMXKVMNBHPAILY-UHFFFAOYSA-N
7800
C14H28O2
MFCD00015065
Dodecanoic acid ethyl ester; Ethyl laurate
CHEBI:87427
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents