missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl DL-Mandelate, TCI America™

$62.64 - $414.03

Chemical Identifiers

CAS 774-40-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00004494
InChI Key SAXHIDRUJXPDOD-UHFFFAOYSA-N
Synonym ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
PubChem CID 13050
ChEBI CHEBI:38750
IUPAC Name ethyl 2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M003925G View Documents TCI America
M003925G
25g Each for $62.64
 
M0039500G View Documents TCI America
M0039500G
500g Each for $414.03
 
Specifications

Chemical Identifiers

774-40-3
180.203
SAXHIDRUJXPDOD-UHFFFAOYSA-N
13050
ethyl 2-hydroxy-2-phenylacetate
C10H12O3
MFCD00004494
ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
CHEBI:38750
CCOC(=O)C(C1=CC=CC=C1)O

Specifications

133°C
25g
C10H12O3
ethyl mandelate, ethyl phenylglycolate, mandelic acid, ethyl ester, dl-mandelic acid ethyl ester, mandelic acid ethyl ester, ethyl dl-mandelate, mandelsaeureaethylester, ethyl hydroxy phenyl acetate, mandelsaeureaethylester german, +-ethyl mandelate
CCOC(=O)C(C1=CC=CC=C1)O
180.203
CHEBI:38750
Crystal-Lump at 20°C
Ethyl DL-Mandelate
White-Yellow
774-40-3
MFCD00004494
SAXHIDRUJXPDOD-UHFFFAOYSA-N
ethyl 2-hydroxy-2-phenylacetate
13050
180.20
≥95.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3841

RTECSNumber : OO7397000

TSCA : No