missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl DL-2-Methylbutyrate, TCI America™

Manufacturer:  TCI America M0757500ML

 View more versions of this product

Catalog No. M0757500ML



Specifications

Specifications

Ethyl DL-2-Methylbutyrate
Colorless
7452-79-1
MFCD00012217
ethyl 2-methylbutyrate, butanoic acid, 2-methyl-, ethyl ester, butyric acid, 2-methyl-, ethyl ester, ethyl 2-methyl butyrate, ethyl alpha-methylbutyrate, dl-2-methylbutyric acid ethyl ester, ethyl 2-methyl-butanoate, ethyl .alpha.-methylbutyrate, 2-methylbutanoic acid ethyl ester, butyric acid, 2-methyl-, ethyl ester 8ci
CCC(C)C(=O)OCC
130.187
130.19
≥98.0% (GC)
133°C
500mL
C7H14O2
3272
HCRBXQFHJMCTLF-UHFFFAOYSA-N
ethyl 2-methylbutanoate
24020
Clear Liquid at 20°C

Chemical Identifiers

7452-79-1
130.187
HCRBXQFHJMCTLF-UHFFFAOYSA-N
24020
CCC(C)C(=O)OCC
C7H14O2
MFCD00012217
ethyl 2-methylbutyrate, butanoic acid, 2-methyl-, ethyl ester, butyric acid, 2-methyl-, ethyl ester, ethyl 2-methyl butyrate, ethyl alpha-methylbutyrate, dl-2-methylbutyric acid ethyl ester, ethyl 2-methyl-butanoate, ethyl .alpha.-methylbutyrate, 2-methylbutanoic acid ethyl ester, butyric acid, 2-methyl-, ethyl ester 8ci
ethyl 2-methylbutanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents