Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl diphenylphosphinite, 98%

$36.03 - $381.74

Chemical Identifiers

Chemical Name or Material Ethyl diphenylphosphinite
CAS 719-80-2
Molecular Formula (C6H5)2POCH2CH3
Formula Weight 230.25
MDL Number MFCD00009087
Beilstein 1641750
Synonym Diphenylphosphinous acid ethyl ester; Ethoxydiphenylphosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1508703

  SDS
alfa aesar™
A1508703
1g Each for $36.03

AAA1508706

  SDS
alfa aesar™
A1508706
5g Each for $91.73

AAA1508714

  SDS
alfa aesar™
A1508714
25g Each for $381.74
Description & Specifications

Specifications

Formula Weight 230.25
Chemical Name or Material Ethyl diphenylphosphinite
Assay 98%
Molecular Formula (C6H5)2POCH2CH3
MDL Number MFCD00009087
CAS 719-80-2
Beilstein 1641750
Synonym Diphenylphosphinous acid ethyl ester; Ethoxydiphenylphosphine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 179° to 180°C (14mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Recommended Storage 2° to 8°C

Applications
Ethyl diphenylphosphinite is used in the synthesis of phosphoramides.

Solubility
Soluble in chloroform.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C