missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl diphenylphosphinite, 98%

Manufacturer:  Alfa Aesar™ A1508714

 View more versions of this product

Catalog No. AAA1508714



Description

Description

Applications
Ethyl diphenylphosphinite is used in the synthesis of phosphoramides.

Solubility
Soluble in chloroform.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Ethyl diphenylphosphinite
179°C to 180°C (14mmHg)
719-80-2
MFCD00009087
ethyl diphenylphosphinite, ethoxydiphenylphosphine, ethyldiphenylphosphinite, phosphinous acid, diphenyl-, ethyl ester, ethyl-diphenylphosphinit, pubchem6468, diphenyethoxy phosphine, ethoxydiphenyl phosphine, acmc-1bbad, ethoxy diphenyl phosphane
CCOP(C1=CC=CC=C1)C2=CC=CC=C2
230.247
230.25
>110°C (230°F)
25g
C14H15OP
1641750
JCRCPEDXAHDCAJ-UHFFFAOYSA-N
ethoxy(diphenyl)phosphane
69754
98%

Chemical Identifiers

719-80-2
230.247
JCRCPEDXAHDCAJ-UHFFFAOYSA-N
69754
CCOP(C1=CC=CC=C1)C2=CC=CC=C2
C14H15OP
MFCD00009087
ethyl diphenylphosphinite, ethoxydiphenylphosphine, ethyldiphenylphosphinite, phosphinous acid, diphenyl-, ethyl ester, ethyl-diphenylphosphinit, pubchem6468, diphenyethoxy phosphine, ethoxydiphenyl phosphine, acmc-1bbad, ethoxy diphenyl phosphane
ethoxy(diphenyl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents