Learn More
Ethyl dimethylpyruvate, 97%, Thermo Scientific Chemicals
$90.75 - $369.90
Chemical Identifiers
CAS | 20201-24-5 |
---|---|
Molecular Formula | C7H12O3 |
Molecular Weight (g/mol) | 144.17 |
MDL Number | MFCD00009122 |
InChI Key | CKTYYUQUWFEUCO-UHFFFAOYSA-N |
Synonym | ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate |
PubChem CID | 88406 |
IUPAC Name | ethyl 3-methyl-2-oxobutanoate |
SMILES | CCOC(=O)C(=O)C(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5341306
|
Thermo Scientific Chemicals
H5341306 |
5 g |
N/A
|
|
|||||
AAH5341314
|
Thermo Scientific Chemicals
H5341314 |
25 g |
N/A
|
|
|||||
Description
Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
20201-24-5 | |
144.17 | |
CKTYYUQUWFEUCO-UHFFFAOYSA-N | |
88406 | |
CCOC(=O)C(=O)C(C)C |
C7H12O3 | |
MFCD00009122 | |
ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate | |
ethyl 3-methyl-2-oxobutanoate |
Specifications
20201-24-5 | |
62°C (11 mmHg) | |
C7H12O3 | |
MFCD00009122 | |
UN3272 | |
ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate | |
CKTYYUQUWFEUCO-UHFFFAOYSA-N | |
ethyl 3-methyl-2-oxobutanoate | |
88406 | |
97% |
0.989 | |
43°C (109°F) | |
1.41 | |
5 g | |
1756668 | |
Slightly soluble in water. | |
CCOC(=O)C(=O)C(C)C | |
144.17 | |
144.17 | |
Ethyl dimethylpyruvate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 243-587-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only