missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl(dimethyl)(2-phenylethyl)ammonium Bis(trifluoromethanesulfonyl)imide, TCI America™

Manufacturer:  TCI America E12831G

 View more versions of this product

Catalog No. E12831G



Specifications

Specifications

Ethyl(dimethyl)(2-phenylethyl)ammonium Bis(trifluoromethanesulfonyl)imide
1g
C14H20F6N2O4S2
XVNMSYLRWLNKMT-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;ethyl-dimethyl-(2-phenylethyl)azanium
121234255
Clear Liquid at 20°C
Yellow
1804970-28-2
Ethyl(dimethyl)phenethylammonium Bis(trifluoromethanesulfonyl)imide
CC[N+](C)(C)CCC1=CC=CC=C1.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
458.434
458.43
≥98.0% (T)

Chemical Identifiers

1804970-28-2
458.434
Ethyl(dimethyl)phenethylammonium Bis(trifluoromethanesulfonyl)imide
bis(trifluoromethylsulfonyl)azanide;ethyl-dimethyl-(2-phenylethyl)azanium
C14H20F6N2O4S2
XVNMSYLRWLNKMT-UHFFFAOYSA-N
121234255
CC[N+](C)(C)CCC1=CC=CC=C1.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents