missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Diformylacetate, TCI America™

Manufacturer:  TCI America E11281G

 View more versions of this product

Catalog No. E11281G



Specifications

Specifications

Ethyl Diformylacetate
Yellow
80370-42-9
MFCD11112084
HMFLBGNCDZYITR-UHFFFAOYSA-N
ethyl 2-formyl-3-oxopropanoate
11029955
Clear Liquid at 20°C
32°C
1g
C6H8O4
Diformylacetic Acid Ethyl Ester, Ethyl 2-Formyl-3-oxopropionate, 2-Formyl-3-oxopropionic Acid Ethyl Ester
CCOC(=O)C(C=O)C=O
144.126
144.13
≥97.0% (T)

Chemical Identifiers

80370-42-9
144.126
HMFLBGNCDZYITR-UHFFFAOYSA-N
11029955
CCOC(=O)C(C=O)C=O
C6H8O4
MFCD11112084
Diformylacetic Acid Ethyl Ester, Ethyl 2-Formyl-3-oxopropionate, 2-Formyl-3-oxopropionic Acid Ethyl Ester
ethyl 2-formyl-3-oxopropanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents