missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl difluoroacetate, 98%

$110.55 - $110.55

Chemical Identifiers

CAS 454-31-9
Molecular Formula F2CHCO2CH2CH3
Formula Weight 124.09
MDL Number MFCD00013578
UN Number UN2924
Beilstein 1748597
Synonym Difluoroacetic acid ethyl ester
Chemical Name or Material Ethyl difluoroacetate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1121506 SDS Alfa Aesar™
A1121506*
5g Each for $110.55
AAA1121522 SDS Alfa Aesar™
A1121522
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1121514 SDS Alfa Aesar™
A1121514
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Ethyl difluoroacetate is used in pharmaceutical intermediates, acrylization oxidation and halogenating reaction of catalyst. Also ethyl difluoroacetate is used in preparing pyrrazolo-pyrimidine derivatives.

Solubility
Slightly soluble in water

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

454-31-9
124.09
UN2924
Difluoroacetic acid ethyl ester
F2CHCO2CH2CH3
MFCD00013578
1748597
Ethyl difluoroacetate

Specifications

5g
98° to 99°C
F2CHCO2CH2CH3
MFCD00013578
1748597
Ethyl difluoroacetate
454-31-9
25°C (77°F)
124.09
UN2924
Difluoroacetic acid ethyl ester
98%

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA