Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl diethoxyphosphinylformate, 98%

$25.62 - $256.15

Chemical Identifiers

Chemical Name or Material Ethyl diethoxyphosphinylformate
CAS 1474-78-8
Molecular Formula C7H15O5P
Formula Weight 210.17
MDL Number MFCD00009108
Beilstein 1781949
Synonym Triethyl phosphonoformate; Diethoxyphosphinylformic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAB2336714

  SDS
alfa aesar™
B2336714
Each for $25.62

AAB2336722

  SDS
alfa aesar™
B2336722
Each for $71.27

AAB2336736

  SDS
alfa aesar™
B2336736
Each for $256.15
Description & Specifications

Specifications

Formula Weight 210.17
Chemical Name or Material Ethyl diethoxyphosphinylformate
TSCA TSCA
Assay 98%
Molecular Formula C7H15O5P
MDL Number MFCD00009108
CAS 1474-78-8
Beilstein 1781949
Synonym Triethyl phosphonoformate; Diethoxyphosphinylformic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 128° to 129°C (9mmHg)
Quantity 25g
Flash Point 119°C (246°F)
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.