Please login to your online account to display your discounted pricing

Ethyl diethoxyacetate ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl diethoxyacetate
Assay Percent Range 98%
Molecular Formula C8H16O4
Linear Formula (C2H5O)2CHCO2C2H5
Formula Weight 176.21
MDL Number MFCD00009231
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC156071000

  SDS
thermo fisher scientific
AC156071000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.21
Quantity 100g
Density 0.989g/mL
Beilstein 03, 601
Fieser 06,253
Chemical Name or Material Ethyl diethoxyacetate
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C8H16O4
Linear Formula (C2H5O)2CHCO2C2H5
MDL Number MFCD00009231
CAS 6065-82-3