Learn More
Ethyl dichlorophosphate, 97%, Thermo Scientific Chemicals
$189.69 - $189.69
Chemical Identifiers
CAS | 1498-51-7 |
---|---|
Molecular Formula | C2H5Cl2O2P |
Molecular Weight (g/mol) | 162.94 |
MDL Number | MFCD00002069 |
InChI Key | YZBOZNXACBQJHI-UHFFFAOYSA-N |
Synonym | ethyl dichlorophosphate, ethyl phosphorodichloridate, phosphorodichloridic acid, ethyl ester, ethyldichlorophosphate, dichloroethoxyphosphine oxide, ethylphosphoric acid dichloride, dichlorophosphoric acid, ethyl ester, unii-4ip5dtw75u, ethylesterdichlorid german, hsdb 417 |
PubChem CID | 15158 |
IUPAC Name | 1-dichlorophosphoryloxyethane |
SMILES | CCOP(=O)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC118380250
|
Thermo Scientific Chemicals
118380250 |
25 g | Glass bottle |
Each for $189.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1498-51-7 | |
162.94 | |
YZBOZNXACBQJHI-UHFFFAOYSA-N | |
15158 | |
CCOP(=O)(Cl)Cl |
C2H5Cl2O2P | |
MFCD00002069 | |
ethyl dichlorophosphate, ethyl phosphorodichloridate, phosphorodichloridic acid, ethyl ester, ethyldichlorophosphate, dichloroethoxyphosphine oxide, ethylphosphoric acid dichloride, dichlorophosphoric acid, ethyl ester, unii-4ip5dtw75u, ethylesterdichlorid german, hsdb 417 | |
1-dichlorophosphoryloxyethane |
Specifications
1498-51-7 | |
1.3730g/mL | |
>110°C | |
97% | |
C2H5Cl2O2P | |
C2H5OP(O)Cl2 | |
25 g | |
1.373 | |
YZBOZNXACBQJHI-UHFFFAOYSA-N | |
1-dichlorophosphoryloxyethane | |
15158 | |
97% | |
Ethyl dichlorophosphate, 97% |
Colorless to Brown | |
167°C (758.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4330 to 1.435 | |
MFCD00002069 | |
01,332 | |
ethyl dichlorophosphate, ethyl phosphorodichloridate, phosphorodichloridic acid, ethyl ester, ethyldichlorophosphate, dichloroethoxyphosphine oxide, ethylphosphoric acid dichloride, dichlorophosphoric acid, ethyl ester, unii-4ip5dtw75u, ethylesterdichlorid german, hsdb 417 | |
CCOP(=O)(Cl)Cl | |
162.94 | |
162.94 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 216-099-0
RTECSNumber : TD4390000
TSCA : TSCA
RUO – Research Use Only