missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl dichlorophosphate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC118380050

 View more versions of this product

Catalog No. AC118380050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl dichlorophosphate
>110°C
Glass bottle
1.373
Colorless to Brown
97%
97%
C2H5OP(O)Cl2
01,332
YZBOZNXACBQJHI-UHFFFAOYSA-N
1-dichlorophosphoryloxyethane
15158
Liquid
1.3730g/mL
Authentic
1.4330 to 1.435
167°C (758.0mmHg)
5g
1498-51-7
C2H5Cl2O2P
MFCD00002069
dichloroethoxyphosphine oxide, dichlorophosphoric acid, ethyl ester, ethyl dichlorophosphate, ethyl phosphorodichloridate, ethyldichlorophosphate, ethylesterdichlorid german, ethylphosphoric acid dichloride, hsdb 417, phosphorodichloridic acid, ethyl ester, unii-4ip5dtw75u
CCOP(=O)(Cl)Cl
162.94
162.94
97%

Chemical Identifiers

1498-51-7
162.94
YZBOZNXACBQJHI-UHFFFAOYSA-N
15158
CCOP(=O)(Cl)Cl
C2H5Cl2O2P
MFCD00002069
dichloroethoxyphosphine oxide, dichlorophosphoric acid, ethyl ester, ethyl dichlorophosphate, ethyl phosphorodichloridate, ethyldichlorophosphate, ethylesterdichlorid german, ethylphosphoric acid dichloride, hsdb 417, phosphorodichloridic acid, ethyl ester, unii-4ip5dtw75u
1-dichlorophosphoryloxyethane
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 216-099-0

RTECSNumber : TD4390000

TSCA : TSCA

SDS Documents