Please login to your online account to display your discounted pricing

Ethyl Cyclopropanecarboxylate 99%, ACROS Organics™

$206.33 - $206.33

Chemical Identifiers

Chemical Name or Material Ethyl cyclopropanecarboxylate
Name Note 99%
CAS 988537
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H10O2
Linear Formula C3H5CO2C2H5
Formula Weight 114.14
MDL Number MFCD00001282
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC118351000

  SDS
thermo fisher scientific
118351000
Glass bottle 100g Each for $206.33
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 114.14
Quantity 100g
Color Clear pale yellow
EINECS Number 225-010-4
Density 96%g/mL
Beilstein 09, 4
Fieser 08,222
CAS 988537
Chemical Name or Material Ethyl cyclopropanecarboxylate
Packaging Glass bottle
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H10O2
Linear Formula C3H5CO2C2H5
MDL Number MFCD00001282
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.