missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Cyclohexylacetate, TCI America™

$94.72 - $902.94

Chemical Identifiers

CAS 5452-75-5
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00001517
InChI Key ZBDAMDWKXGTKBT-UHFFFAOYSA-N
Synonym ethyl cyclohexylacetate, ethyl cyclohexaneacetate, cyclohexaneacetic acid, ethyl ester, ethyl-2-cyclohexylacetate, ethylcyclohexylacetate, unii-v36o028g9r, cyclohexylacetic acid ethyl ester, cyclohexaneacetic acid ethyl ester, cyclohexyl acetic acid ethyl ester, pubchem20245
PubChem CID 21595
IUPAC Name ethyl 2-cyclohexylacetate
SMILES CCOC(=O)CC1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C107625ML View Documents TCI America
C107625ML
25mL Each for $94.72
 
C1076500ML View Documents TCI America
C1076500ML
500mL Each for $902.94
 
Specifications

Chemical Identifiers

5452-75-5
170.252
ZBDAMDWKXGTKBT-UHFFFAOYSA-N
21595
CCOC(=O)CC1CCCCC1
C10H18O2
MFCD00001517
ethyl cyclohexylacetate, ethyl cyclohexaneacetate, cyclohexaneacetic acid, ethyl ester, ethyl-2-cyclohexylacetate, ethylcyclohexylacetate, unii-v36o028g9r, cyclohexylacetic acid ethyl ester, cyclohexaneacetic acid ethyl ester, cyclohexyl acetic acid ethyl ester, pubchem20245
ethyl 2-cyclohexylacetate

Specifications

212°C
25mL
C10H18O2
ethyl cyclohexylacetate, ethyl cyclohexaneacetate, cyclohexaneacetic acid, ethyl ester, ethyl-2-cyclohexylacetate, ethylcyclohexylacetate, unii-v36o028g9r, cyclohexylacetic acid ethyl ester, cyclohexaneacetic acid ethyl ester, cyclohexyl acetic acid ethyl ester, pubchem20245
CCOC(=O)CC1CCCCC1
170.252
170.25
≥98.0% (GC)
Colorless
5452-75-5
MFCD00001517
ZBDAMDWKXGTKBT-UHFFFAOYSA-N
ethyl 2-cyclohexylacetate
21595
Clear Liquid at 20°C
Ethyl Cyclohexylacetate
Safety and Handling

Safety and Handling

RTECSNumber : GU6555000

TSCA : Yes