Please login to your online account to display your discounted pricing

Ethyl Cyclohexylacetate +98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  118340500

 View more versions of this product

Catalog No. AC118340500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 0.9400g/mL
Packaging Glass bottle
Quantity 50g
Chemical Name or Material Ethyl cyclohexylacetate
Name Note 98+%
CAS 5452-75-5
Assay Percent Range 98+%
Molecular Formula C10H18O2
Linear Formula C6H11CH2CO2C2H5
Formula Weight 170.25g/mol
MDL Number MFCD00001517
Beilstein 09, 14
Synonym Ethyl cyclohexaneacetate
RTECS Number GU6555000
EINECS Number 226-695-2
TSCA TSCA

Specifications

Chemical Name or Material Ethyl cyclohexylacetate
Name Note 98+%
CAS 5452-75-5
Assay Percent Range 98+%
Molecular Formula C10H18O2
Linear Formula C6H11CH2CO2C2H5
Formula Weight 170.25g/mol
MDL Number MFCD00001517
Synonym Ethyl cyclohexaneacetate