Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl cyclohexylacetate, 98+%

$32.64 - $32.64

Chemical Identifiers

Chemical Name or Material Ethyl cyclohexylacetate
CAS 5452-75-5
Formula Weight 170.25g/mol
Synonym Cyclohexylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2466306

 
alfa aesar™
B24663-06
5g Each for $32.64
Description & Specifications

Specifications

Boiling Point 211° to 212°C
Flash Point 80°C (176°F)
Quantity 5g
Chemical Name or Material Ethyl cyclohexylacetate
CAS 5452-75-5
Formula Weight 170.25g/mol
Synonym Cyclohexylacetic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.