missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl cyclohexanecarboxylate, 98+%, Thermo Scientific Chemicals
$157.55 - $565.92
Chemical Identifiers
CAS | 3289-28-9 |
---|---|
Molecular Formula | C9H16O2 |
Molecular Weight (g/mol) | 156.225 |
MDL Number | MFCD00041734 |
InChI Key | JJOYCHKVKWDMEA-UHFFFAOYSA-N |
Synonym | cyclohexanecarboxylic acid, ethyl ester, ethyl cyclohexylformate, ethyl hexahydrobenzoate, cyclohexanecarboxylic acid ethyl ester, ethoxycarbonylcyclohexane, ethyl cyclohexylcarboxylate, ethyl cyclohexanoate, fema no. 3544, ethyl cyclohexylmethanoate, ethyl cyclohexanocarboxylate |
PubChem CID | 18686 |
IUPAC Name | ethyl cyclohexanecarboxylate |
SMILES | CCOC(=O)C1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0262206
|
Thermo Scientific Chemicals
L0262206 |
5 g |
Each for $157.55
|
|
|||||
AAL0262214
|
Thermo Scientific Chemicals
L0262214 |
25 g |
Each for $565.92
|
|
|||||
Description
Ethyl cyclohexanecarboxylate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl cyclohexanecarboxylate is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Chemical Identifiers
3289-28-9 | |
156.225 | |
JJOYCHKVKWDMEA-UHFFFAOYSA-N | |
18686 | |
CCOC(=O)C1CCCCC1 |
C9H16O2 | |
MFCD00041734 | |
cyclohexanecarboxylic acid, ethyl ester, ethyl cyclohexylformate, ethyl hexahydrobenzoate, cyclohexanecarboxylic acid ethyl ester, ethoxycarbonylcyclohexane, ethyl cyclohexylcarboxylate, ethyl cyclohexanoate, fema no. 3544, ethyl cyclohexylmethanoate, ethyl cyclohexanocarboxylate | |
ethyl cyclohexanecarboxylate |
Specifications
3289-28-9 | |
194°C to 196°C | |
1.442 | |
5 g | |
cyclohexanecarboxylic acid, ethyl ester, ethyl cyclohexylformate, ethyl hexahydrobenzoate, cyclohexanecarboxylic acid ethyl ester, ethoxycarbonylcyclohexane, ethyl cyclohexylcarboxylate, ethyl cyclohexanoate, fema no. 3544, ethyl cyclohexylmethanoate, ethyl cyclohexanocarboxylate | |
JJOYCHKVKWDMEA-UHFFFAOYSA-N | |
ethyl cyclohexanecarboxylate | |
18686 | |
≥98% |
0.936 | |
C9H16O2 | |
MFCD00041734 | |
2042698 | |
Insoluble in water. | |
CCOC(=O)C1CCCCC1 | |
156.225 | |
156.23 | |
Ethyl cyclohexanecarboxylate |
Safety and Handling
EINECSNumber : 221-945-7
RTECSNumber : GU8490000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only