missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl cycloheptylideneacetate, 97%

$28.53 - $85.14

Chemical Identifiers

CAS 1903-23-7
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD02093941
InChI Key RAWVIWMURMZEQF-UHFFFAOYSA-N
Synonym ethyl cycloheptylideneacetate, ethyl 2-1e-cycloheptylidene acetate, acetic acid, 2-cycloheptylidene-, ethyl ester, cycloheptylideneacetic acid ethyl ester, cycloheptylidene-acetic acid ethyl ester, acetic acid,2-cycloheptylidene-, ethyl ester
PubChem CID 2757986
IUPAC Name ethyl 2-cycloheptylideneacetate
SMILES CCOC(=O)C=C1CCCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1760603 View Documents Alfa Aesar™
L1760603
1g Each for $28.53
 
AAL1760606 View Documents Alfa Aesar™
L1760606
5g Each for $85.14
 
Specifications

Chemical Identifiers

1903-23-7
182.263
RAWVIWMURMZEQF-UHFFFAOYSA-N
2757986
CCOC(=O)C=C1CCCCCC1
C11H18O2
MFCD02093941
ethyl cycloheptylideneacetate, ethyl 2-1e-cycloheptylidene acetate, acetic acid, 2-cycloheptylidene-, ethyl ester, cycloheptylideneacetic acid ethyl ester, cycloheptylidene-acetic acid ethyl ester, acetic acid,2-cycloheptylidene-, ethyl ester
ethyl 2-cycloheptylideneacetate

Specifications

115°C to 116°C (10mmHg)
1903-23-7
MFCD02093941
ethyl cycloheptylideneacetate, ethyl 2-1e-cycloheptylidene acetate, acetic acid, 2-cycloheptylidene-, ethyl ester, cycloheptylideneacetic acid ethyl ester, cycloheptylidene-acetic acid ethyl ester, acetic acid,2-cycloheptylidene-, ethyl ester
CCOC(=O)C=C1CCCCCC1
182.263
182.26
Ethyl cycloheptylideneacetate
1g
C11H18O2
2088922
RAWVIWMURMZEQF-UHFFFAOYSA-N
ethyl 2-cycloheptylideneacetate
2757986
97%