missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl cycloheptylideneacetate, 97%

Manufacturer:  Alfa Aesar™ L1760603

 View more versions of this product

Catalog No. AAL1760603



Specifications

Specifications

Ethyl cycloheptylideneacetate
1g
C11H18O2
2088922
RAWVIWMURMZEQF-UHFFFAOYSA-N
ethyl 2-cycloheptylideneacetate
2757986
97%
115°C to 116°C (10mmHg)
1903-23-7
MFCD02093941
acetic acid, 2-cycloheptylidene-, ethyl ester
CCOC(=O)C=C1CCCCCC1
182.263
182.26

Chemical Identifiers

1903-23-7
182.263
RAWVIWMURMZEQF-UHFFFAOYSA-N
2757986
CCOC(=O)C=C1CCCCCC1
C11H18O2
MFCD02093941
acetic acid, 2-cycloheptylidene-, ethyl ester
ethyl 2-cycloheptylideneacetate
Documents