Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl cycloheptylacetate, 98%

$38.86 - $38.86

Chemical Identifiers

Chemical Name or Material Ethyl cycloheptylacetate
CAS 80246-70-4
Molecular Formula C11H20O2
Formula Weight 184.28
MDL Number MFCD08437072
Synonym Cycloheptylacetic acid ethyl ester; Ethyl cycloheptaneacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2586603

  SDS
alfa aesar™
H2586603
1g Each for $38.86
Description & Specifications

Specifications

Formula Weight 184.28
Molecular Formula C11H20O2
Quantity 1g
Assay 98%
Boiling Point 80° to 85°C (2mmHg)
CAS 80246-70-4
Synonym Cycloheptylacetic acid ethyl ester; Ethyl cycloheptaneacetate
MDL Number MFCD08437072
Chemical Name or Material Ethyl cycloheptylacetate