missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Coumarin-3-carboxylate, TCI America™

Manufacturer:  TCI America E09925G

 View more versions of this product

Catalog No. E09925G



Specifications

Specifications

Ethyl Coumarin-3-carboxylate
94°C
1846-76-0
MFCD00016964
XKHPEMKBJGUYCM-UHFFFAOYSA-N
ethyl 2-oxochromene-3-carboxylate
15800
Crystal-Powder at 20°C
White
5g
C12H10O4
ethyl coumarin-3-carboxylate, ethyl 2-oxo-2h-chromene-3-carboxylate, 3-ethoxycarbonylcoumarin, ethyl 3-coumarincarboxylate, 3-carbethoxycoumarin, ethylcoumarin-3-carboxylate, coumarin-3-carboxylic acid ethyl ester, 1,2-benzopyran-2-one-3-carboxylic acid, ethyl ester, 2h-1-benzopyran-3-carboxylic acid, 2-oxo-, ethyl ester, 2h-1-benzopyran-3-carboxylicacid, 2-oxo-, ethyl ester
CCOC(=O)C1=CC2=CC=CC=C2OC1=O
218.208
218.21
≥98.0% (GC)

Chemical Identifiers

1846-76-0
218.208
XKHPEMKBJGUYCM-UHFFFAOYSA-N
15800
CCOC(=O)C1=CC2=CC=CC=C2OC1=O
C12H10O4
MFCD00016964
ethyl coumarin-3-carboxylate, ethyl 2-oxo-2h-chromene-3-carboxylate, 3-ethoxycarbonylcoumarin, ethyl 3-coumarincarboxylate, 3-carbethoxycoumarin, ethylcoumarin-3-carboxylate, coumarin-3-carboxylic acid ethyl ester, 1,2-benzopyran-2-one-3-carboxylic acid, ethyl ester, 2h-1-benzopyran-3-carboxylic acid, 2-oxo-, ethyl ester, 2h-1-benzopyran-3-carboxylicacid, 2-oxo-, ethyl ester
ethyl 2-oxochromene-3-carboxylate
Safety and Handling

Safety and Handling

RTECSNumber : DJ2501000

TSCA : No

Documents