missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 265640025
Specifications
Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride | |
Glass bottle | |
128°C to 135°C | |
95% | |
95% | |
MFCD00144280 | |
Solubility in water: soluble in water. Other solubilities: soluble in methanol | |
CCOC(=O)C1CCCCC1N.Cl | |
207.7 | |
207.7 | |
95% |
Authentic | |
White | |
2.5g | |
1127-99-7 | |
C9H17NO2·HCl | |
1r,2s-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cis-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, cis-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cyclohexanecarboxylic acid, 2-amino-, ethyl ester, hydrochloride, cis, ethyl 1r,2s-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl 2s,1r-2-aminocyclohexanecarboxylate, chloride, ethyl cis-2-amino-1-cyclohexanecarboxylate hcl salt, ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, ethyl cis-2-aminocyclohexanecarboxylate hydrochloride | |
XMQSOBPCWYVZSW-WLYNEOFISA-N | |
ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate;hydrochloride | |
2724641 | |
Crystalline Powder or Chunks |
Chemical Identifiers
1127-99-7 | |
207.7 | |
XMQSOBPCWYVZSW-WLYNEOFISA-N | |
2724641 | |
CCOC(=O)C1CCCCC1N.Cl |
C9H17NO2·HCl | |
MFCD00144280 | |
1r,2s-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cis-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, cis-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cyclohexanecarboxylic acid, 2-amino-, ethyl ester, hydrochloride, cis, ethyl 1r,2s-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl 2s,1r-2-aminocyclohexanecarboxylate, chloride, ethyl cis-2-amino-1-cyclohexanecarboxylate hcl salt, ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, ethyl cis-2-aminocyclohexanecarboxylate hydrochloride | |
ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate;hydrochloride |
Safety and Handling
MOISTURE SENSITIVE