missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 265640010

 View more versions of this product

Catalog No. AC265640010



Description

Description

Specifications

Specifications

Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride
Glass bottle
128°C to 135°C
95%
95%
MFCD00144280
Solubility in water: soluble in water. Other solubilities: soluble in methanol
CCOC(=O)C1CCCCC1N.Cl
207.7
207.7
95%
Authentic
White
1g
1127-99-7
C9H17NO2·HCl
1r,2s-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cis-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, cis-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cyclohexanecarboxylic acid, 2-amino-, ethyl ester, hydrochloride, cis, ethyl 1r,2s-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl 2s,1r-2-aminocyclohexanecarboxylate, chloride, ethyl cis-2-amino-1-cyclohexanecarboxylate hcl salt, ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, ethyl cis-2-aminocyclohexanecarboxylate hydrochloride
XMQSOBPCWYVZSW-WLYNEOFISA-N
ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate;hydrochloride
2724641
Crystalline Powder or Chunks

Chemical Identifiers

1127-99-7
207.7
XMQSOBPCWYVZSW-WLYNEOFISA-N
2724641
CCOC(=O)C1CCCCC1N.Cl
C9H17NO2·HCl
MFCD00144280
1r,2s-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cis-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, cis-ethyl 2-aminocyclohexanecarboxylate hydrochloride, cyclohexanecarboxylic acid, 2-amino-, ethyl ester, hydrochloride, cis, ethyl 1r,2s-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl 2s,1r-2-aminocyclohexanecarboxylate, chloride, ethyl cis-2-amino-1-cyclohexanecarboxylate hcl salt, ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, ethyl cis-2-aminocyclohexanecarboxylate hydrochloride
ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate;hydrochloride
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

SDS Documents