missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Cinnamate 99.0+%, TCI America™
Supplier: TCI America C0359100G
Specifications
| Ethyl Cinnamate | |
| 10°C | |
| 271°C | |
| MFCD00009189 | |
| ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate | |
| CCOC(=O)C=CC1=CC=CC=C1 | |
| 176.215 | |
| CHEBI:4895 | |
| ≥99.0% (GC) |
| 103-36-6 | |
| Yellow | |
| C11H12O2 | |
| 100 g | |
| KBEBGUQPQBELIU-CMDGGOBGSA-N | |
| ethyl (E)-3-phenylprop-2-enoate | |
| 637758 | |
| 176.22 | |
| Liquid |
Chemical Identifiers
| 103-36-6 | |
| 176.215 | |
| KBEBGUQPQBELIU-CMDGGOBGSA-N | |
| 637758 | |
| ethyl (E)-3-phenylprop-2-enoate |
| C11H12O2 | |
| MFCD00009189 | |
| ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate | |
| CHEBI:4895 | |
| CCOC(=O)C=CC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-1750
RTECSNumber : GD9010000
TSCA : Yes