Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl chrysanthemate, 95%

Chemical Identifiers

Chemical Name or Material Ethyl chrysanthemate
CAS 97-41-6
Formula Weight 196.29
MDL Number MFCD00001304
Beilstein 2045740
Synonym Ethyl 2,2-dimethyl-3-(1-isobutenyl)cyclopropane-1-carboxylate; Chrysanthemum monocarboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5958914

  SDS
alfa aesar™
H5958914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5958922

  SDS
alfa aesar™
H5958922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.29
Chemical Name or Material Ethyl chrysanthemate
TSCA TSCA
Refractive Index 1.4615
Color Yellow
Boiling Point 112°C (10mmHg)
Physical Form Liquid
Assay 95%
MDL Number MFCD00001304
CAS 97-41-6
Beilstein 2045740
Synonym Ethyl 2,2-dimethyl-3-(1-isobutenyl)cyclopropane-1-carboxylate; Chrysanthemum monocarboxylic acid ethyl ester
Density 0.906g/mL
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GZ1727000
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.