missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl chrysanthemate, 95%

Manufacturer:  Alfa Aesar™ H5958922

 View more versions of this product

Catalog No. AAH5958922



Specifications

Specifications

Ethyl chrysanthemate
1.4615
Yellow
97-41-6
MFCD00001304
ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate
CCOC(=O)C1C(C1(C)C)C=C(C)C
196.29
196.29
95%
0.906g/mL
112°C (10mmHg)
100g
C12H20O2
2045740
VIMXTGUGWLAOFZ-UHFFFAOYSA-N
ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
7334
Liquid

Chemical Identifiers

97-41-6
196.29
VIMXTGUGWLAOFZ-UHFFFAOYSA-N
7334
CCOC(=O)C1C(C1(C)C)C=C(C)C
C12H20O2
MFCD00001304
ethyl chrysanthemate, ethyl chrysanthemumate, chrysanthemic acid ethyl ester, cyclopropanecarboxylic acid, 2,2-dimethyl-3-2-methyl-1-propenyl-, ethyl ester, ethylchrysanthemate, ccris 2498, chrysanthemic acid, ethyl ester, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methylprop-1-en-1-yl cyclopropanecarboxylate, ethyl 2,2-dimethyl-3-2-methyl-1-propenyl cyclopropane-1-carboxylate
ethyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GZ1727000

TSCA : TSCA

SDS Documents