Please login to your online account to display your discounted pricing

Ethyl chlorothioformate, 96%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  347160010

 View more versions of this product

Catalog No. AC347160010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4815 to 1.4835 (20°C, 589nm)
Color Clear yellow or gray-green to purple
Quantity 1g
Chemical Name or Material Ethyl chlorothioformate
Name Note 96%
CAS 2941-64-2
Physical Form Clear yellow or gray-greenish to purple liquid
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C3H5ClOS
Linear Formula ClCOSC2H5
Formula Weight 124.58g/mol
MDL Number MFCD00037110
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number FG3855000
EINECS Number 220-928-1
TSCA TSCA

Specifications

Chemical Name or Material Ethyl chlorothioformate
Name Note 96%
CAS 2941-64-2
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C3H5ClOS
Linear Formula ClCOSC2H5
Formula Weight 124.58g/mol
MDL Number MFCD00037110
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents