Please login to your online account to display your discounted pricing

Ethyl Chloroformate 99%, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC118250025

 View more versions of this product

Catalog No. AC118250025

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay 98.5% max. (GC) 0.3% max. (ethanol) (GC) 1.0% max. (diethyl carbonate) (GC)
Formula Weight 108.52
Quantity 2.5L
Color Clear
EINECS Number 208-778-5
TSCA TSCA
Density 1.1350g/mL
RTECS Number LQ6125000
Beilstein 03, 10
Fieser 01,364; 02,193; 04,228; 05,294; 07,147; 09,214; 12,223
Merck Index 15, 3839
Phosgene phosgene: < 0.1%
Iron (Fe) 5ppm max.
Packaging Glass bottle
Carbonate 1% max.
Alcohol alcohol: < 0.3%
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
Free Acid 0.1 % max. (HCl)
Chemical Name or Material Ethyl chloroformate
Infrared Spectrum Authentic
Refractive Index 1.3940 to 1.3960 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C3H5ClO2
Linear Formula ClCO2C2H5
MDL Number MFCD00000644
CAS 541-41-3
Synonym Ethyl chlorocarbonate
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ethyl chloroformate
Name Note 99%
CAS 541-41-3
Assay 98.5% max. (GC) 0.3% max. (ethanol) (GC) 1.0% max. (diethyl carbonate) (GC)
Assay Percent Range 99%
Molecular Formula C3H5ClO2
Linear Formula ClCO2C2H5
Formula Weight 108.52
MDL Number MFCD00000644
Synonym Ethyl chlorocarbonate
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.

SDS & Safety Documents