Learn More
Ethyl chloroacetate, 99%, Thermo Scientific Chemicals
$44.22 - $189.10
Chemical Identifiers
CAS | 105-39-5 |
---|---|
Molecular Formula | C4H7ClO2 |
Molecular Weight (g/mol) | 122.548 |
MDL Number | MFCD00000932 |
InChI Key | VEUUMBGHMNQHGO-UHFFFAOYSA-N |
Synonym | ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf |
PubChem CID | 7751 |
IUPAC Name | ethyl 2-chloroacetate |
SMILES | CCOC(=O)CCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1555422
|
Thermo Scientific Chemicals
A1555422 |
100 g |
Each for $44.22
|
|
|||||
AAA1555436
|
Thermo Scientific Chemicals
A1555436 |
500 g |
Each for $73.70
|
|
|||||
AAA155540E
|
Thermo Scientific Chemicals
A155540E |
2500 g |
Each for $189.10
|
|
|||||
Description
Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.
Solubility
Soluble in water 20 g/L (20°C).
Notes
Incompatible with acids, bases, oxidizing agents, reducing agents.
Chemical Identifiers
105-39-5 | |
122.548 | |
VEUUMBGHMNQHGO-UHFFFAOYSA-N | |
7751 | |
CCOC(=O)CCl |
C4H7ClO2 | |
MFCD00000932 | |
ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf | |
ethyl 2-chloroacetate |
Specifications
105-39-5 | |
1.159 | |
54°C (149°F) | |
C4H7ClO2 | |
ClCH2CO2CH2CH3 | |
100 g | |
506455 | |
ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf | |
VEUUMBGHMNQHGO-UHFFFAOYSA-N | |
ethyl 2-chloroacetate | |
7751 | |
99% |
-26°C | |
142°C to 144°C | |
Pungent | |
1.421 | |
MFCD00000932 | |
UN1181 | |
14,3783 | |
Soluble in water 20g/L (20°C). | |
CCOC(=O)CCl | |
122.548 | |
122.55 | |
Ethyl chloroacetate |
Safety and Handling
GHS H Statement
H301-H311-H331-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P333+P313-P363-P370+P378q-P501c
H226-H300-H311+H331-H315-H317-H318
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ETHYL CHLOROACETATE
EINECSNumber : 203-294-0
RTECSNumber : AF9110000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only