Please login to your online account to display your discounted pricing

Ethyl cellulose, ethoxyl content 48%, 4 cps, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  190925000

 View more versions of this product

Catalog No. AC190925000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Granular Powder
Quantity 500g
Color White to light yellow
TSCA TSCA
Assay 48 to 49.5% (ethoxyl)
Viscosity 3 to 5.5cPs (5% soln. in 80/20 toluene/ethanol,25°C)
RTECS Number FJ5950500
Merck Index 15, 3836
Chemical Name or Material Ethyl cellulose
MDL Number MFCD00131037
CAS 9004-57-3
Infrared Spectrum Authentic
Packaging Plastic bottle

Specifications

Chemical Name or Material Ethyl cellulose
Assay 48 to 49.5% (ethoxyl)
MDL Number MFCD00131037