Please login to your online account to display your discounted pricing

Ethyl cellulose, ethoxyl content 48%, 4 cps, ACROS Organics™

$426.42 - $426.42

Chemical Identifiers

Chemical Name or Material Ethyl cellulose
Assay 48 to 49.5% (ethoxyl)
MDL Number MFCD00131037
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC190925000

  SDS
thermo fisher scientific
190925000
Plastic bottle 500g Each for $426.42
Description & Specifications

Specifications

Physical Form Granular Powder
Quantity 500g
Color White to light yellow
Assay 48 to 49.5% (ethoxyl)
TSCA TSCA
RTECS Number FJ5950500
Viscosity 3 to 5.5cPs (5% soln. in 80/20 toluene/ethanol,25°C)
Merck Index 15, 3836
Infrared Spectrum Authentic
Packaging Plastic bottle
MDL Number MFCD00131037
Chemical Name or Material Ethyl cellulose
CAS 9004-57-3