missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Cellulose [9-11mPa.s, 5% in Toluene + Ethanol (80:20) at 25°C], TCI America™

Manufacturer:  TCI America E0265500G

 View more versions of this product

Catalog No. E0265500G



Specifications

Specifications

Ethyl Cellulose [9-11mPa.s, 5% in Toluene + Ethanol (80:20) at 25°C]
500g
C23H24N6O4
n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene
CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
448.483
Crystal-Powder at 20°C
White-Yellow
9004-57-3
MFCD00131037
ARSKJXYLLONUAJ-UHFFFAOYSA-N
2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate
166304

Chemical Identifiers

9004-57-3
448.483
ARSKJXYLLONUAJ-UHFFFAOYSA-N
166304
CCCCN(CCOC(=O)C)C1=CC(=C(C=C1)N=NC2=C(C=C(C=C2C#N)[N+](=O)[O-])C#N)C
C23H24N6O4
MFCD00131037
n-butyl-n-2-acetoxyethyl-4-4-nitro-2,6-dicyanophenyl azo-3-methylbenzeneamine, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-4-2-acetyloxy ethyl butylamino-2-methylphenyl azo-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, 1,3-benzenedicarbonitrile, 2-2-4-2-acetyloxy ethyl butylamino-2-methylphenyl diazenyl-5-nitro, azo dye n5, 4-2,6-dicyano-4-nitrophenyl azo-3-methylphenyl amino hexyl acetate, ethyl cellulose, viscosity ethoxyl, 2-n-butyl-4-2,6-dicyano-4-nitrophenyl diazenyl-3-methylanilino ethyl acetate, 4-n-2-acetoxyethyl butyl amino-2',6'-dicyano-2-methyl-4'-nitroazobenzene
2-[N-butyl-4-[(2,6-dicyano-4-nitrophenyl)diazenyl]-3-methylanilino]ethyl acetate
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0186

RTECSNumber : FJ5950500

TSCA : Yes

Documents