Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl cellulose

$87.54 - $87.54

Chemical Identifiers

Chemical Name or Material Ethyl cellulose
CAS 9004-57-3
MDL Number MFCD00131037
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27621MD

  SDS
alfa aesar™
H27621MD
250mg Each for $87.54
Description & Specifications

Specifications

Chemical Name or Material Ethyl cellulose
TSCA TSCA
Quantity 250mg
MDL Number MFCD00131037
CAS 9004-57-3
RTECS Number FJ5950500

Applications
Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color. In addition to this, it is used in lacquers and plastic toughing.

Solubility
Soluble in tetrahydrofuran, methyl acetate, ethanol, methanol, toluene, ethyl acetate, chloroform, esters, aromatic hydrocarbons, alcohols and ketones. Insoluble in water, glycerol and propane-1,2-diol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and strong bases.