missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl carbamate, 98%

Manufacturer:  Alfa Aesar™ 4480418

 View more versions of this product

Catalog No. AA4480418



Specifications

Specifications

Ethyl carbamate
182°C to 184°C
48°C to 50°C
51-79-6
H2NCO2C2H5
635810
urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
CCOC(=O)N
89.094
CHEBI:17967
Crystalline
>92°C (198°F)
White
50g
C3H7NO2
MFCD00007966
14,9877
JOYRKODLDBILNP-UHFFFAOYSA-N
ethyl carbamate
5641
89.09
98%

Chemical Identifiers

51-79-6
89.094
JOYRKODLDBILNP-UHFFFAOYSA-N
5641
ethyl carbamate
C3H7NO2
MFCD00007966
urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
CHEBI:17967
CCOC(=O)N
Safety and Handling

Safety and Handling

GHS H Statement
H350
May cause cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : FA8400000

TSCA : TSCA

SDS Documents