missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Caprylate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150510050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl caprylate | |
106-32-1 | |
-47°C | |
0.8680g/mL | |
75°C | |
99% min. (GC) | |
C10H20O2 | |
CH3(CH2)6CO2C2H5 | |
5 mL | |
0.868 | |
ethyl caprylate, octanoic acid, ethyl ester, ethyl n-octanoate, caprylic acid ethyl ester, ethyl octoate, ethyl octylate, octanoic acid ethyl ester, natural, unii-81c5mop582, fema no. 2449 | |
YYZUSRORWSJGET-UHFFFAOYSA-N | |
ethyl octanoate | |
7799 | |
172.27 | |
Liquid |
99+% | |
1mg KOH/g max. | |
Colorless | |
206°C to 208°C | |
Authentic | |
Glass bottle | |
1.4150 to 1.42 | |
MFCD00009552 | |
02, 348 | |
15, 3833 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether,insoluble in glycerol | |
CCCCCCCC(=O)OCC | |
172.27 | |
CHEBI:87426 | |
99+% |
Chemical Identifiers
106-32-1 | |
172.27 | |
YYZUSRORWSJGET-UHFFFAOYSA-N | |
7799 | |
ethyl octanoate |
C10H20O2 | |
MFCD00009552 | |
ethyl caprylate, octanoic acid, ethyl ester, ethyl n-octanoate, caprylic acid ethyl ester, ethyl octoate, ethyl octylate, octanoic acid ethyl ester, natural, unii-81c5mop582, fema no. 2449 | |
CHEBI:87426 | |
CCCCCCCC(=O)OCC |
Safety and Handling
EINECSNumber : 203-385-5
RTECSNumber : RH0680000
TSCA : TSCA
RUO – Research Use Only