missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl caprate, 99+%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 165000050

 View more versions of this product

Catalog No. AC165000050



Specifications

Specifications

1mg KOH/g max.
0.8665g/mL
Authentic
1.4240 to 1.4270
<30 mPa.s (20°C)
245.0°C
-20.0°C
99+%
99% min. (GC)
CH3(CH2)8CO2C2H5
02,356
capric acid ethyl ester
RGXWDWUGBIJHDO-UHFFFAOYSA-N
ethyl decanoate
8048
200.32
99+%
Ethyl caprate
102°C
Glass bottle
0.8665
0.3% max. (K.F.)
Colorless
5mL
110-38-3
C12H24O2
MFCD00009581
15,3831
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
CCCCCCCCCC(=O)OCC
200.32
CHEBI:87430
Liquid

Chemical Identifiers

110-38-3
200.32
RGXWDWUGBIJHDO-UHFFFAOYSA-N
8048
ethyl decanoate
C12H24O2
MFCD00009581
capric acid ethyl ester
CHEBI:87430
CCCCCCCCCC(=O)OCC
Safety and Handling

Safety and Handling

EINECSNumber : 203-761-9

RTECSNumber : HD9420000

TSCA : TSCA

SDS Documents