missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl butyrylacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1278618
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl butyrylacetate | |
-44°C | |
94°C to 95°C (12 mmHg) | |
C8H14O3 | |
MFCD00009401 | |
507689 | |
KQWWVLVLVYYYDT-UHFFFAOYSA-N | |
ethyl 3-oxohexanoate | |
238498 | |
158.2 |
3249-68-1 | |
0.99 | |
78°C (172°F) | |
1.427 | |
50 g | |
ethyl butyrylacetate, ethyl butyroacetate, hexanoic acid, 3-oxo-, ethyl ester, ethyl beta-ketohexanoate, ethyl alpha-butyrylacetate, 3-ketohexanoic acid ethyl ester, ethyl-3-oxohexanoate, unii-8q1ahg710e, ethyl .alpha.-butyrylacetate, fema no. 3683 | |
CCCC(=O)CC(=O)OCC | |
158.20 | |
CHEBI:18119 | |
98% |
Chemical Identifiers
3249-68-1 | |
158.20 | |
KQWWVLVLVYYYDT-UHFFFAOYSA-N | |
238498 | |
ethyl 3-oxohexanoate |
C8H14O3 | |
MFCD00009401 | |
ethyl butyrylacetate, ethyl butyroacetate, hexanoic acid, 3-oxo-, ethyl ester, ethyl beta-ketohexanoate, ethyl alpha-butyrylacetate, 3-ketohexanoic acid ethyl ester, ethyl-3-oxohexanoate, unii-8q1ahg710e, ethyl .alpha.-butyrylacetate, fema no. 3683 | |
CHEBI:18119 | |
CCCC(=O)CC(=O)OCC |
Safety and Handling
GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
H227-H302
EINECSNumber : 221-835-9
RTECSNumber : MO8420500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only