missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl butyrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 118182500

 View more versions of this product

Catalog No. AC118182500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl butyrate
105-54-4
-93.3°C
0.8700g/mL
26°C
98.5% min. (GC)
C6H12O2
CH3CH2CH2CO2C2H5
250 mL
0.87
ethyl butyrate, butyric acid ethyl ester, ethyl n-butyrate, butanoic acid, ethyl ester, butyric ester, butyric ether, butyric acid, ethyl ester, ethyl n-butanoate, butanoic acid ethyl ester, fema number 2427
OBNCKNCVKJNDBV-UHFFFAOYSA-N
ethyl butanoate
7762
99%
99%
1mg KOH/g max.
Colorless
120.0°C
Authentic
Glass bottle
1.3910 to 1.3930
MFCD00009394
02, 270
15, 3830
Solubility in water: practically insoluble
CCCC(=O)OCC
116.16
116.16
Liquid

Chemical Identifiers

105-54-4
116.16
OBNCKNCVKJNDBV-UHFFFAOYSA-N
7762
CCCC(=O)OCC
C6H12O2
MFCD00009394
ethyl butyrate, butyric acid ethyl ester, ethyl n-butyrate, butanoic acid, ethyl ester, butyric ester, butyric ether, butyric acid, ethyl ester, ethyl n-butanoate, butanoic acid ethyl ester, fema number 2427
ethyl butanoate
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.

GHS P Statement
Wear eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep cool.

GHS Signal Word: Warning

EINECSNumber : 203-306-4

RTECSNumber : ET1660000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only